D,L-Nicotine hydrochloride, [pyrrolidine-2-14C] ≥97% (by HPLC)

Supplier: MORAVEK BIOCHEMICALS MS

MC484-10UCI MC484-50UCI
101972-008EA 1693.46 USD
101972-008 101972-010
D,L-Nicotine hydrochloride, [pyrrolidine-2-14C] ≥97% (by HPLC)
D,L-Nicotine hydrochloride, [pyrrolidine-2-14C]
CAS Number: 337511-53-2
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR