Benzylamine hydrchloride, [7-14C] ≥97% (by HPLC)

Supplier: MORAVEK BIOCHEMICALS MS

MC468-50UCI MC468-250UCI
101971-964EA 5387.69 USD
101971-964 101971-966
Benzylamine hydrchloride, [7-14C] ≥97% (by HPLC)
Benzylamine hydrchloride, [7-14C]
CAS Number: 71367-20-9
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR