Arachidonyl coenzyme A, [arachidonyl-1-14C] ≥96%

Supplier: MORAVEK BIOCHEMICALS MS

MC459-10UCI MC459-50UCI
101971-942EA 2040 USD
101971-942 101971-944
Arachidonyl coenzyme A, [arachidonyl-1-14C] ≥96%
Arachidonyl coenzyme A, [arachidonyl-1-14C]
CAS Number: 289887-63-4
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR