1-(2-Deoxy-2-fluoroarabinofuranosyl)-uracil, [uracil-2-14C] ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC439-10UCI MC439-50UCI
101971-896EA 1693.46 USD
101971-896 101971-898
1-(2-Deoxy-2-fluoroarabinofuranosyl)-uracil, [uracil-2-14C] ≥98% (by HPLC), MORpure™
1-(2-Deoxy-2-fluoroarabinofuranosyl)-uracil, [uracil-2-14C]
CAS Number: 114988-21-5
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR