Oleic acid, [1-14C] ≥97% (by HPLC)

Supplier: MORAVEK BIOCHEMICALS MS

MC406-50UCI MC406-250UCI
101971-846EA 5387.69 USD
101971-846 101971-848
Oleic acid, [1-14C] ≥97% (by HPLC)
Oleic acid, [1-14C]
CAS Number: 3343-81-5
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR