L-Leucine, [1-14C] ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC404-50UCI MC404-250UCI MC404-1MCI
101971-834EA 5387.69 USD
101971-834 101971-836 101971-838
L-Leucine, [1-14C] ≥98%, MORpure™
L-Leucine, [1-14C]
CAS Number: 6109-63-3
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR