p-Aminobenzoic acid, [carboxyl-14C] ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC400-50UCI MC400-250UCI MC400-1MCI
101971-824EA 5387.69 USD
101971-824 101971-826 101971-828
p-Aminobenzoic acid, [carboxyl-14C] ≥98% (by HPLC), MORpure™
p-Aminobenzoic acid, [carboxyl-14C]
CAS Number: 21482-31-5
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR