Citric acid, [1,5-14C] ≥97%

Supplier: MORAVEK BIOCHEMICALS MS

MC365-50UCI MC365-250UCI
101971-820EA 5387.69 USD
101971-820 101971-822
Citric acid, [1,5-14C] ≥97%
Citric acid, [1,5-14C]
CAS Number: 58308-53-5
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR