Arachidonic acid, [1-14C] ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC364-10UCI MC364-50UCI MC364-250UCI
101971-814EA 1693.46 USD
101971-814 101971-816 101971-818
Arachidonic acid, [1-14C] ≥98%, MORpure™
Arachidonic acid, [1-14C]
CAS Number: 3435-81-2
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR