2-Deoxy-D-glucose, [1-14C] ≥97%

Supplier: MORAVEK BIOCHEMICALS MS

MC355S-50UCI MC355S-250UCI MC355S-1MCI
101971-802EA 5191.54 USD
101971-802 101971-804 101971-806
2-Deoxy-D-glucose, [1-14C] ≥97%
2-Deoxy-D-glucose, [1-14C]
CAS Number: 72561-26-3
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR