Butyric acid, sodium salt, [1-14C] ≥97% (by HPLC)

Supplier: MORAVEK BIOCHEMICALS MS

MC319-50UCI MC319-250UCI MC319-1MCI
101971-786EA 5387.69 USD
101971-786 101971-788 101971-790
Butyric acid, sodium salt, [1-14C] ≥97% (by HPLC)
Butyric acid, sodium salt, [1-14C]
CAS Number: 62327-34-8
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR