5-Iodouracil, [2-14C] ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC315-10UCI MC315-50UCI
101971-780EA 1693.46 USD
101971-780 101971-782
5-Iodouracil, [2-14C] ≥98%, MORpure™
5-Iodouracil, [2-14C]
CAS Number: 26149-71-3
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR