Adefovir, [8-14C] ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC303-10UCI MC303-50UCI
101971-750EA 1693.46 USD
101971-750 101971-752
Adefovir, [8-14C] ≥98%, MORpure™
Adefovir, [8-14C]
CAS Number: 145074-48-2
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR