Benzoic acid, [7-14C] ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC290-100UCI MC290-250UCI MC290-500UCI MC290-1MCI MC290-5MCI
101971-706EA 5544.62 USD
101971-706 101971-708 101971-710 101971-712 101971-714
Benzoic acid, [7-14C] ≥98%, MORpure™
Benzoic acid, [7-14C]
CAS Number: 1589-66-8
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR