L-Valine, [14C(U)] ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC277-50UCI MC277-250UCI
101971-686EA 5387.69 USD
101971-686 101971-688
L-Valine, [14C(U)] ≥98% (by HPLC), MORpure™
L-Valine, [14C(U)]
CAS Number: 4303-77-9
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR