L-Tyrosine, [14C(U)] ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC275-50UCI MC275-250UCI MC275-1MCI
101971-676EA 5387.69 USD
101971-676 101971-678 101971-680
L-Tyrosine, [14C(U)] ≥98%, MORpure™
L-Tyrosine, [14C(U)]
CAS Number: 4367-87-7
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR