L-Proline, [14C(U)] ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC263-50UCI MC263-250UCI
101971-636EA 5387.69 USD
101971-636 101971-638
L-Proline, [14C(U)] ≥98%, MORpure™
L-Proline, [14C(U)]
CAS Number: 2728-13-4
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR