L-Phenylalanine, [14C(U)] ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC262-50UCI MC262-250UCI
101971-632EA 5387.69 USD
101971-632 101971-634
L-Phenylalanine, [14C(U)] ≥98%, MORpure™
L-Phenylalanine, [14C(U)]
CAS Number: 1465783-91-8
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR