Sodium cyanide, [14C] ≥97% (by HPLC)

Supplier: MORAVEK BIOCHEMICALS MS

MC256-5MCI MC256-25MCI
101971-628EA 5446.54 USD
101971-628 101971-630
Sodium cyanide, [14C] ≥97% (by HPLC)
Sodium cyanide, [14C]
CAS Number: 3396-82-5
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR