Potassium thiocyanate, [14C] ≥97%

Supplier: MORAVEK BIOCHEMICALS MS

MC243-250UCI MC243-1MCI
101971-600EA 4646.23 USD
101971-600 101971-602
Potassium thiocyanate, [14C] ≥97%
Potassium thiocyanate, [14C]
CAS Number: 38527-79-6
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR