Zebularine, [2-14C] ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC242-10UCI MC242-50UCI
101971-588EA 1693.46 USD
101971-588 101971-590
Zebularine, [2-14C] ≥98% (by HPLC), MORpure™
Zebularine, [2-14C]
CAS Number: 34340-05-1
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR