Uracil-β-D-arabinofuranoside, [2-14C] ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC241-10UCI MC241-50UCI
101971-564EA 1693.46 USD
101971-564 101971-566
Uracil-β-D-arabinofuranoside, [2-14C] ≥98%, MORpure™
Uracil-β-D-arabinofuranoside, [2-14C]
CAS Number: 336804-66-1
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR