Prochloraz, [phenyl-14C(U)] ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC2331-10UCI MC2331-50UCI
101971-418EA 1693.46 USD
101971-418 101971-420
Prochloraz, [phenyl-14C(U)] ≥98%, MORpure™
Prochloraz, [phenyl-14C(U)]
CAS Number: 2088204-01-5
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR