3,4-Diaminopyridine phosphate, [2,6-14C] ≥97% (by HPLC)

Supplier: MORAVEK BIOCHEMICALS MS

MC2325-10UCI MC2325-50UCI
101971-406EA 1693.46 USD
101971-406 101971-408
3,4-Diaminopyridine phosphate, [2,6-14C] ≥97% (by HPLC)
3,4-Diaminopyridine phosphate, [2,6-14C]
CAS Number: 2088858-51-7
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR