Uridine, [uracil-14C(U)] ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC2313-10UCI MC2313-50UCI
101971-386EA 1693.46 USD
101971-386 101971-388
Uridine, [uracil-14C(U)] ≥98%, MORpure™
Uridine, [uracil-14C(U)]
CAS Number: 2088858-43-7
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR