Thymidine, [2,6-14C] ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC2304-10UCI MC2304-50UCI
101971-370EA 1693.46 USD
101971-370 101971-372
Thymidine, [2,6-14C] ≥98%, MORpure™
Thymidine, [2,6-14C]
CAS Number: 2089095-53-2
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR