Cyanoacetic acid, [cyano-14C] ≥97%

Supplier: MORAVEK BIOCHEMICALS MS

MC225-250UCI MC225-1MCI
101971-266EA 6394.62 USD
101971-266 101971-268
Cyanoacetic acid, [cyano-14C] ≥97%
Cyanoacetic acid, [cyano-14C]
CAS Number: 58454-80-1
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR