Decanoyl coenzyme A, [decanoic acid-1-14C] ≥95%

Supplier: MORAVEK BIOCHEMICALS MS

MC2206-10UCI MC2206-50UCI
101971-170EA 2183.85 USD
101971-170 101971-172
Decanoyl coenzyme A, [decanoic acid-1-14C] ≥95%
Decanoyl coenzyme A, [decanoic acid-1-14C]
CAS Number: 288608-43-5
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR