Naphthalene, [1,4,5,8-14C] ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC2147-50UCI MC2147-250UCI
101971-076EA 5387.69 USD
101971-076 101971-078
Naphthalene, [1,4,5,8-14C] ≥98%, MORpure™
Naphthalene, [1,4,5,8-14C]
CAS Number: 503298-00-8
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR