4-Aminobutyric acid, [4-14C] ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC2145-250UCI MC2145-50UCI
101971-070EA 7414.62 USD
101971-070 101971-068
4-Aminobutyric acid, [4-14C] ≥98%, MORpure™
4-Aminobutyric acid, [4-14C]
CAS Number: 21195-45-9
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR