Thymidine-5'-triphosphate, tetraammonium salt, [2-14C] ≥95% (by HPLC)

Supplier: MORAVEK BIOCHEMICALS MS

MC2142-10UCI MC2142-50UCI
101971-056EA 1693.46 USD
101971-056 101971-058
Thymidine-5'-triphosphate, tetraammonium salt, [2-14C] ≥95% (by HPLC)
Thymidine-5'-triphosphate, tetraammonium salt, [2-14C]
CAS Number: 2088577-42-6
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR