Acetamidine hydrochloride, [1-14C] ≥97%

Supplier: MORAVEK BIOCHEMICALS MS

MC2073-50UCI MC2073-250UCI MC2073-1MCI
101971-002EA 5387.69 USD
101971-002 101971-004 101971-006
Acetamidine hydrochloride, [1-14C] ≥97%
Acetamidine hydrochloride, [1-14C]
CAS Number: 847666-56-2
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR