D-Mannosamine, [1-14C] ≥97% (by HPLC)

Supplier: MORAVEK BIOCHEMICALS MS

MC2063-50UCI MC2063-250UCI
101970-994EA 5387.69 USD
101970-994 101970-996
D-Mannosamine, [1-14C] ≥97% (by HPLC)
D-Mannosamine, [1-14C]
CAS Number: 289623-56-9
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR