2-Pyrrolidone, [5-14C] ≥97%

Supplier: MORAVEK BIOCHEMICALS MS

MC2058-50UCI MC2058-250UCI
101970-980EA 5387.69 USD
101970-980 101970-982
2-Pyrrolidone, [5-14C] ≥97%
2-Pyrrolidone, [5-14C]
CAS Number: 50494-55-8
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR