Metformin hydrochloride, [biguanidine-14C] ≥96%

Supplier: MORAVEK BIOCHEMICALS MS

MC2043-50UCI MC2043-250UCI MC2043-1MCI
101970-966EA 1791.54 USD
101970-966 101970-968 101970-970
Metformin hydrochloride, [biguanidine-14C] ≥96%
Metformin hydrochloride, [biguanidine-14C]
CAS Number: 1703804-73-2
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR