Creatine hydrate, [4-14C] ≥97%

Supplier: MORAVEK BIOCHEMICALS MS

MC204-50UCI MC204-250UCI
101970-960EA 5387.69 USD
101970-960 101970-962
Creatine hydrate, [4-14C] ≥97%
Creatine hydrate, [4-14C]
CAS Number: 336804-77-4
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR