2,4-Diamino-6-nitrotoluene, [methyl-14C] ≥95% (by HPLC)

Supplier: MORAVEK BIOCHEMICALS MS

MC2037-10UCI MC2037-50UCI
101970-948EA 1693.46 USD
101970-948 101970-950
2,4-Diamino-6-nitrotoluene, [methyl-14C] ≥95% (by HPLC)
2,4-Diamino-6-nitrotoluene, [methyl-14C]
CAS Number: 2088139-12-0
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR