Isobutyric acid, sodium salt, [1-14C] ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC2022-50UCI MC2022-250UCI
101970-906EA 5387.69 USD
101970-906 101970-908
Isobutyric acid, sodium salt, [1-14C] ≥98% (by HPLC), MORpure™
Isobutyric acid, sodium salt, [1-14C]
CAS Number: 6917-21-1
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR