D-Mannose, [1-14C] ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC202-50UCI MC202-250UCI
101970-898EA 5387.69 USD
101970-898 101970-900
D-Mannose, [1-14C] ≥98%, MORpure™
D-Mannose, [1-14C]
CAS Number: 4314-62-9
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR