Propionic acid, sodium salt, [1-14C] ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC201-50UCI MC201-250UCI MC201-1MCI
101970-876EA 5387.69 USD
101970-876 101970-878 101970-880
Propionic acid, sodium salt, [1-14C] ≥98% (by HPLC), MORpure™
Propionic acid, sodium salt, [1-14C]
CAS Number: 4561-17-5
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR