2-Chloroadenosine, [8-14C] ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC198-10UCI MC198-50UCI
101970-856EA 1693.46 USD
101970-856 101970-858
2-Chloroadenosine, [8-14C] ≥98%, MORpure™
2-Chloroadenosine, [8-14C]
CAS Number: 336804-75-2
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR