Bromoacetic acid, [1-14C] ≥97% (by HPLC)

Supplier: MORAVEK BIOCHEMICALS MS

MC188-50UCI MC188-250UCI MC188-1MCI
101970-826EA 5191.54 USD
101970-826 101970-828 101970-830
Bromoacetic acid, [1-14C] ≥97% (by HPLC)
Bromoacetic acid, [1-14C]
CAS Number: 4561-21-1
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR