Xanthosine-5'-monophosphate, diammonium salt, [8-14C] ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC182-10UCI MC182-50UCI
101970-812EA 1693.46 USD
101970-812 101970-814
Xanthosine-5'-monophosphate, diammonium salt, [8-14C] ≥98% (by HPLC), MORpure™
Xanthosine-5'-monophosphate, diammonium salt, [8-14C]
CAS Number: 335021-78-8
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR