Xanthine, [8-14C] ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC181-50UCI MC181-250UCI
101970-804EA 5387.69 USD
101970-804 101970-806
Xanthine, [8-14C] ≥98%, MORpure™
Xanthine, [8-14C]
CAS Number: 23199-23-7
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR