Uridine-5'-monophosphate, diammonium salt, [2-14C] ≥97% (by HPLC)

Supplier: MORAVEK BIOCHEMICALS MS

MC180-10UCI MC180-25UCI MC180-50UCI
101970-798EA 1693.46 USD
101970-798 101970-800 101970-802
Uridine-5'-monophosphate, diammonium salt, [2-14C] ≥97% (by HPLC)
Uridine-5'-monophosphate, diammonium salt, [2-14C]
CAS Number: 288245-90-9
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR