Xanthosine, [8-14C] ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC178-10UCI MC178-50UCI
101970-790EA 1693.46 USD
101970-790 101970-792
Xanthosine, [8-14C] ≥98% (by HPLC), MORpure™
Xanthosine, [8-14C]
CAS Number: 2777-90-4
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR