Guanosine, [8-14C] ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC177-10UCI MC177-50UCI
101970-784EA 1693.46 USD
101970-784 101970-786
Guanosine, [8-14C] ≥98% (by HPLC), MORpure™
Guanosine, [8-14C]
CAS Number: 129356-67-8
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR