L-Leucine, [14C(U)] ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC175-50UCI MC175-250UCI
101970-770EA 1373.08 USD
101970-770 101970-772 101970-774
L-Leucine, [14C(U)] ≥98%, MORpure™
L-Leucine, [14C(U)]
CAS Number: 5155-21-5
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR