Formic acid, sodium salt, [14C] ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC173-50UCI MC173-250UCI MC173-1MCI MC173-5MCI
101970-758EA 5387.69 USD
101970-758 101970-760 101970-762 101970-764
Formic acid, sodium salt, [14C] ≥98% (by HPLC), MORpure™
Formic acid, sodium salt, [14C]
CAS Number: 2792-70-3
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR