Uracil, [6-14C] ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC159-50UCI MC159-250UCI
101970-722EA 5387.69 USD
101970-722 101970-724
Uracil, [6-14C] ≥98%, MORpure™
Uracil, [6-14C]
CAS Number: 58454-83-4
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR